Assay Details
Target Gene Details
Entrez Gene ID: | 60533 |
Gene Name: | CD274 antigen |
Gene Aliases: |
A530045L16Rik, B7h1, Pdcd1l1, Pdcd1lg1, Pdl1 |
Location: |
Chr.19:29367438-29388095 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd274 | NM_021893.3 | 3 | 367 | NP_068693.1 |
| AF233517.1 | 2 | 300 | AAG18509.1 | |
| AF317088.1 | 2 | 284 | AAG31810.1 | |
| AK154403.1 | 3 | 346 | BAE32562.1 | |
| BC066841.1 | 3 | 350 | AAH66841.1 | |
| BY193530.1 | ||||
| GQ904196.1 | 2 | 284 |
More Information
Set Membership: |
|

Genomic Map