Assay Details
Target Gene Details
Entrez Gene ID: | 60533 |
Gene Name: | CD274 antigen |
Gene Aliases: |
A530045L16Rik, B7h1, Pdcd1l1, Pdcd1lg1, Pdl1 |
Location: |
Chr.19:29367438-29388095 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd274 | NM_021893.3 | 7 | 1383 | NP_068693.1 |
| AF233517.1 | 6 | 1316 | AAG18509.1 | |
| AK154403.1 | 7 | 1362 | BAE32562.1 | |
| BC066841.1 | 7 | 1366 | AAH66841.1 |
More Information
Set Membership: |
|

Genomic Map