Assay Details
Target Gene Details
Entrez Gene ID: | 209091 |
Gene Name: | cyclin B3 |
Gene Aliases: |
- |
Location: |
Chr.X:6979652-7041635 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb3 | NM_183015.3 | 7 | 1200 | NP_898836.2 |
| AJ555464.1 | 7 | 1201 | CAD88194.1 | |
| AJ555465.1 | CAD88195.1 | |||
| BC132411.1 | 6 | 1154 | ||
| BC138355.1 | 6 | 1154 |
More Information
Set Membership: |
|

Genomic Map