Assay Details
Target Gene Details
Entrez Gene ID: | 26370 |
Gene Name: | centrin 2 |
Gene Aliases: |
1110034A02Rik, AI326150, Calt, caltractin |
Location: |
Chr.X:72913565-72918344 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cetn2 | NM_019405.6 | 5 | 825 | NP_062278.2 |
| AK004081.1 | 5 | 826 | BAB23161.1 | |
| AK010541.1 | 5 | 816 | BAB27017.1 | |
| AK088488.1 | 3 | 657 | ||
| AK140156.1 | 5 | 843 | BAE24259.1 | |
| AK151033.1 | 5 | 893 | BAE30050.1 | |
| BC013545.1 | 4 | 809 | AAH13545.1 |
More Information
Set Membership: |
|

Genomic Map