Assay Details
Target Gene Details
Entrez Gene ID: | 67298 |
Gene Name: | G protein-coupled receptor associated sorting protein 1 |
Gene Aliases: |
2210415K24Rik, 3110031O14Rik, C87852, GASP, GASP1 |
Location: |
Chr.X:135742692-135803468 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprasp1 | NM_001004359.2 | 10 | 4209 | NP_001004359.1 |
| NM_001005385.1 | 10 | 4095 | NP_001005385.1 | |
| NM_026081.5 | 11 | 4298 | NP_080357.4 | |
| AB093241.1 | 5 | 3415 | BAC41425.1 | |
| AK039156.1 | BAC30259.1 | |||
| BC027187.1 | 1 | 1537 | AAH27187.1 | |
| BC058970.1 | AAH58970.1 | |||
| BC085157.1 | 10 | 3997 | AAH85157.1 |
More Information
Set Membership: |
|

Genomic Map