Assay Details
Target Gene Details
Entrez Gene ID: | 70237 |
Gene Name: | basic helix-loop-helix domain containing, class B9 |
Gene Aliases: |
2700087I09Rik |
Location: |
Chr.X:135841639-135891081 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhb9 | NM_001098222.1 | 5 | 2041 | NP_001091692.1 |
| NM_198161.2 | 5 | 2051 | NP_937804.1 | |
| AK012577.1 | 1 | 534 | ||
| AK214737.1 | 1 | 113 | ||
| BC059871.1 | 1 | 1406 | AAH59871.1 |
More Information
Set Membership: |
|

Genomic Map