Assay Details
Target Gene Details
Entrez Gene ID: | 76308 |
Gene Name: | RAB1B, member RAS oncogene family |
Gene Aliases: |
1110011F09Rik |
Location: |
Chr.19:5099207-5106996 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab1b | NM_029576.3 | 6 | 1614 | NP_083852.1 |
| AK003609.1 | 6 | 1615 | BAB22888.1 | |
| AK042020.1 | 6 | 1616 | BAE20616.1 | |
| AK156726.1 | 6 | 1637 | BAE33821.1 | |
| AK205715.1 | 1 | 177 | ||
| AK211449.1 | 1 | 239 | ||
| AK212653.1 | 1 | 109 | ||
| AK214573.1 | 1 | 148 | ||
| AK215051.1 | 1 | 178 | ||
| AK215541.1 | 1 | 239 | ||
| BC016408.1 | 6 | 1609 | AAH16408.1 |

Genomic Map