Assay Details
Target Gene Details
Entrez Gene ID: | 70237 |
Gene Name: | basic helix-loop-helix domain containing, class B9 |
Gene Aliases: |
2700087I09Rik |
Location: |
Chr.X:135841639-135891081 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhb9 |
Target Gene Details
Entrez Gene ID: | 245607 |
Gene Name: | G protein-coupled receptor associated sorting protein 2 |
Gene Aliases: |
5330440H13Rik, GASP-2, GASP2, Prpl5 |
Location: |
Chr.X:135839034-135844730 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprasp2 | NM_001163015.1 | 7 | 2234 | NP_001156487.1 |
| NM_001163016.1 | 7 | 2196 | NP_001156488.1 | |
| NM_001163017.1 | 7 | 2181 | NP_001156489.1 | |
| AK030638.1 | 7 | 2175 | ||
| BC117923.2 | 7 | 2200 | ||
| BC117924.2 | 7 | 2150 |
More Information
Set Membership: |
|

Genomic Map