Assay Details
Target Gene Details
Entrez Gene ID: | 53623 |
Gene Name: | glutamate receptor, ionotropic, AMPA3 (alpha 3) |
Gene Aliases: |
2900064I19Rik, GluA3, GluR-C, GluR-K3, Glur-3, Glur3, Gluralpha3 |
Location: |
Chr.X:41401119-41678601 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria3 | NM_001281929.1 | 1 | 283 | NP_001268858.1 |
| NM_001290451.1 | 1 | 55 | NP_001277380.1 | |
| NM_016886.4 | 1 | 283 | NP_058582.3 | |
| NR_104053.1 | 1 | 283 | ||
| AB022342.1 | 1 | 73 | BAA37124.1 | |
| AB079072.1 | BAE06153.1 | |||
| AB102775.1 | ||||
| AB102776.1 | ||||
| AF483494.1 | AAL90768.1 | |||
| AF483495.1 | AAL90769.1 | |||
| AK135043.1 | BAE22396.1 | |||
| AK138615.1 | 1 | 188 | ||
| AK157815.1 | 2 | 460 | BAE34208.1 | |
| AK161163.1 | 1 | 103 | BAE36222.1 | |
| AK220530.1 | 1 | 70 | BAD90527.1 | |
| BC086678.1 | 1 | 228 | ||
| BC129855.1 | 1 | 73 | ||
| BC129856.1 | 1 | 73 | ||
| BY266507.1 | 1 | 283 | ||
| CJ107988.1 | 1 | 281 |
More Information
Set Membership: |
|

Genomic Map