Assay Details
Target Gene Details
Entrez Gene ID: | 53623 |
Gene Name: | glutamate receptor, ionotropic, AMPA3 (alpha 3) |
Gene Aliases: |
2900064I19Rik, GluA3, GluR-C, GluR-K3, Glur-3, Glur3, Gluralpha3 |
Location: |
Chr.X:41401119-41678601 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria3 | NM_001281929.1 | 16 | 4071 | NP_001268858.1 |
| NM_016886.4 | 16 | 4060 | NP_058582.3 | |
| NR_104053.1 | 17 | 4175 | ||
| AK220530.1 | 16 | 3858 | BAD90527.1 | |
| BC086678.1 | 16 | 4005 |
More Information
Set Membership: |
|

Genomic Map