Assay Details
Target Gene Details
Entrez Gene ID: | 68505 |
Gene Name: | vacuolar protein sorting 51 homolog (S. cerevisiae) |
Gene Aliases: |
1110014N23Rik, 3110057M17Rik, AI837850, Ffr |
Location: |
Chr.19:6067842-6077187 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps51 | NM_001081041.1 | 10 | 2271 | NP_001074510.1 |
| AK003717.1 | 1 | 440 | ||
| AK137155.1 | 10 | 2281 | BAE23255.1 | |
| AK146211.1 | 10 | 2280 | BAE26981.1 | |
| AK180223.1 | 3 | 370 | ||
| AK195187.1 | 3 | 288 | ||
| AK212543.1 | 2 | 250 | ||
| AK216884.1 | 3 | 288 |

Genomic Map