Assay Details
Target Gene Details
Entrez Gene ID: | 19290 |
Gene Name: | purine rich element binding protein A |
Gene Aliases: |
CAGER-1, Pur-alpha, ssCRE-BP |
Location: |
Chr.18:36280551-36291721 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pura | NM_008989.3 | 2 | 1194 | NP_033015.1 |
| AF017631.1 | 1 | 719 | AAB71860.1 | |
| AK075821.1 | 2 | 863 | BAC35987.1 | |
| U02098.1 | 2 | 1194 | AAA64630.1 |
More Information
Set Membership: |
|

Genomic Map