Assay Details
Target Gene Details
Entrez Gene ID: | 226180 |
Gene Name: | internexin neuronal intermediate filament protein, alpha |
Gene Aliases: |
AV028420, NF-66, NF66 |
Location: |
Chr.19:47014698-47024358 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ina | NM_146100.4 | 3 | 1587 | NP_666212.3 |
| AK144917.1 | 3 | 1034 | BAE26134.1 | |
| BC018383.1 | 3 | 1587 | AAH18383.1 | |
| L36390.1 | 3 | 1583 | AAB37740.1 |

Genomic Map