Assay Details
Target Gene Details
Entrez Gene ID: | 27356 |
Gene Name: | insulin-like 6 |
Gene Aliases: |
RIF1 |
Location: |
Chr.19:29321354-29325318 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Insl6 | NM_013754.1 | 2 | 404 | NP_038782.1 |
| AF135823.1 | 2 | 401 | ||
| AF143807.2 | 2 | 404 | AAD30158.2 | |
| AK005622.1 | 2 | 441 | BAB24156.1 | |
| BC028752.1 | 2 | 416 | AAH28752.1 |
More Information
Set Membership: |
|

Genomic Map