Assay Details
Target Gene Details
Entrez Gene ID: | 14933 |
Gene Name: | glycerol kinase |
Gene Aliases: |
D930012N15Rik, GK |
Location: |
Chr.X:85701937-85776819 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gyk | NM_001294138.1 | 20 | 4016 | NP_001281067.1 |
| NM_001294140.1 | 21 | 4037 | NP_001281069.1 | |
| NM_008194.3 | 19 | 3932 | NP_032220.1 | |
| NM_212444.2 | 20 | 4019 | NP_997609.1 | |
| AK037128.1 | 1 | 908 | ||
| BQ176629.1 | 1 | 426 |
More Information
Set Membership: |
|

Genomic Map