Assay Details
Target Gene Details
Entrez Gene ID: | 70415 |
Gene Name: | serine/threonine kinase 26 |
Gene Aliases: |
2610018G03Rik, Mst4 |
Location: |
Chr.X:50829694-50893098 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk26 | NM_133729.1 | 12 | 1542 | NP_598490.1 |
| AK029278.1 | 10 | 2147 | ||
| AK034609.1 | 12 | 1581 | BAC28768.1 | |
| BC005708.1 | 12 | 1542 | AAH05708.1 | |
| BC132438.1 | ||||
| BC138193.1 |

Genomic Map