Assay Details
Target Gene Details
Entrez Gene ID: | 319262 |
Gene Name: | FCH and double SH3 domains 1 |
Gene Aliases: |
A030002D08Rik |
Location: |
Chr.18:37957434-37969814 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fchsd1 | NM_175684.4 | 20 | 3075 | NP_783615.2 |
| AK154732.1 | 20 | 3110 | ||
| BC057367.1 | 20 | 3075 | AAH57367.1 |
Target Gene Details
Entrez Gene ID: | 225392 |
Gene Name: | RELT-like 2 |
Gene Aliases: |
4631403P03Rik, ependolin |
Location: |
Chr.18:37955084-37959179 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rell2 | NM_153793.2 | NP_722488.2 | ||
| AK014510.1 | 1 | 171 | ||
| AK044097.1 | BAC31774.1 | |||
| BC038500.1 | 6 | 1426 | AAH38500.1 |

Genomic Map