Assay Details
Target Gene Details
Entrez Gene ID: | 59057 |
Gene Name: | zinc finger protein 191 |
Gene Aliases: |
3526401F17Rik, 5033419P20Rik, AI480505, AI642085, AL024422, KOX17, ZF-12, Znf191, Znf24, Znp191 |
Location: |
Chr.18:24009717-24022219 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp191 | NM_021559.2 | 4 | 2323 | NP_067534.2 |
| AK076173.1 | 1 | 1046 | ||
| AK088914.1 | 1 | 1356 | BAC40649.1 | |
| AK146726.1 | 4 | 2321 | BAE27389.1 | |
| AY832930.1 | 4 | 2362 | AAV91152.1 | |
| BC054832.1 | 4 | 2323 | AAH54832.1 |
More Information
Set Membership: |
|

Genomic Map