Assay Details
Target Gene Details
Entrez Gene ID: | 22694 |
Gene Name: | zinc finger protein 35 |
Gene Aliases: |
Zfp-35, Znf271, mszf13, mszf20-1, mszf28, mszf68 |
Location: |
Chr.18:23989634-24005371 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp35 | NM_011755.2 | 3 | 2129 | NP_035885.2 |
| AK132703.1 | 3 | 2132 | BAE21311.1 | |
| AK159080.1 | 3 | 2083 | BAE34798.1 | |
| BC138830.1 | 3 | 2048 | ||
| BC138832.1 | 2 | 2046 | ||
| X17617.1 | 3 | 2080 | CAA35618.1 |
More Information
Set Membership: |
|

Genomic Map