Assay Details
Target Gene Details
Entrez Gene ID: | 16452 |
Gene Name: | Janus kinase 2 |
Gene Aliases: |
Fd17 |
Location: |
Chr.19:29251776-29313080 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jak2 | NM_001048177.2 | 1 | 263 | NP_001041642.1 |
| NM_008413.3 | 1 | 263 | NP_032439.2 | |
| AK087255.1 | 1 | 228 | ||
| BE981299.1 | 1 | 68 | ||
| CJ046359.1 | 1 | 264 | ||
| CJ056249.1 | 1 | 239 |

Genomic Map