Assay Details
Target Gene Details
Entrez Gene ID: | 332396 |
Gene Name: | potassium channel, subfamily K, member 18 |
Gene Aliases: |
Gm781, Tresk, Tresk-2, Trik |
Location: |
Chr.19:59219648-59237370 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk18 | NM_207261.3 | 3 | 1742 | NP_997144.1 |
| AK139600.1 | 3 | 2176 | BAE24080.1 |
More Information
Set Membership: |
|

Genomic Map