Assay Details
Target Gene Details
Entrez Gene ID: | 19671 |
Gene Name: | RCE1 homolog, prenyl protein peptidase (S. cerevisiae) |
Gene Aliases: |
D19Ertd283e, D19Ertd98e |
Location: |
Chr.19:4622551-4625617 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rce1 | NM_023131.1 | 8 | 1215 | NP_075620.1 |
| AK012946.1 | 8 | 1171 | BAB28566.1 | |
| BC004060.1 | 7 | 1064 | AAH04060.1 |
More Information
Set Membership: |
|

Genomic Map