Assay Details
Target Gene Details
Entrez Gene ID: | 104383 |
Gene Name: | REST corepressor 2 |
Gene Aliases: |
1A13, AW122124, CoREST, Rcor, Rcor1 |
Location: |
Chr.19:7267330-7275225 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcor2 | NM_054048.3 | 11 | 1876 | NP_473389.1 |
| AK052295.1 | 13 | 2130 | BAC34922.1 | |
| AK205817.1 | 1 | 232 | ||
| BC055719.1 | 12 | 2297 | AAH55719.1 | |
| X83587.2 | 11 | 1878 | CAB93943.1 |

Genomic Map