Assay Details
Target Gene Details
Entrez Gene ID: | 19697 |
Gene Name: | v-rel reticuloendotheliosis viral oncogene homolog A (avian) |
Gene Aliases: |
p65 |
Location: |
Chr.19:5637440-5648130 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rela | NM_009045.4 | 1 | 172 | NP_033071.1 |
| AK155321.1 | 1 | 182 | ||
| AK158072.1 | 1 | 182 | BAE34348.1 | |
| BC094053.1 | 1 | 172 | AAH94053.1 |
More Information
Set Membership: |
|

Genomic Map