Assay Details
Target Gene Details
Entrez Gene ID: | 19697 |
Gene Name: | v-rel reticuloendotheliosis viral oncogene homolog A (avian) |
Gene Aliases: |
p65 |
Location: |
Chr.19:5637440-5648130 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rela | NM_009045.4 | 11 | 1824 | NP_033071.1 |
| AK089489.1 | 11 | 1569 | BAC40904.1 | |
| AK144565.1 | 11 | 1570 | ||
| AK154630.1 | 11 | 1569 | BAE32727.1 | |
| AK155321.1 | 11 | 1833 | ||
| AK155889.1 | 11 | 1566 | BAE33485.1 | |
| AK157076.1 | 11 | 1556 | BAE33954.1 | |
| AK158072.1 | 11 | 1834 | BAE34348.1 | |
| AK212853.1 | 1 | 242 | ||
| BC003818.1 | 11 | 1583 | AAH03818.1 | |
| BC094053.1 | 11 | 1824 | AAH94053.1 | |
| DQ020177.1 | 9 | 1393 | AAY44745.1 | |
| M61909.1 | 11 | 1539 | AAA39811.1 |
More Information
Set Membership: |
|

Genomic Map