Assay Details
Target Gene Details
Entrez Gene ID: | 331392 |
Gene Name: | nucleolar and coiled-body phosphoprotein 1 pseudogene |
Gene Aliases: |
EG331392 |
Location: |
Chr.X:21360865-21364627 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5124 | NR_045668.1 | 1 | 696 | |
| AK032684.1 | 1 | 697 | BAC27986.1 | |
| BC137827.1 | 1 | 677 | ||
| BM943806.1 | 1 | 611 |

Genomic Map