Assay Details
Target Gene Details
Entrez Gene ID: | 331392 |
Gene Name: | nucleolar and coiled-body phosphoprotein 1 pseudogene |
Gene Aliases: |
EG331392 |
Location: |
Chr.X:21360865-21364627 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5124 | NR_045668.1 | 2 | 1769 | |
| AI426008.1 | 1 | 207 | ||
| AK032684.1 | 2 | 1770 | BAC27986.1 | |
| BC137827.1 | 2 | 1867 |

Genomic Map