Assay Details
Target Gene Details
Entrez Gene ID: | 19877 |
Gene Name: | Rho-associated coiled-coil containing protein kinase 1 |
Gene Aliases: |
1110055K06Rik, Rock-I |
Location: |
Chr.18:10064401-10181792 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rock1 | NM_009071.2 | 33 | 5098 | NP_033097.1 |
| AK004265.1 | 1 | 80 | ||
| AK050269.1 | 9 | 1679 | BAC34154.1 | |
| AK077297.1 | 1 | 448 | ||
| AK171291.1 | 13 | 2123 | BAE42373.1 |
More Information
Set Membership: |
|

Genomic Map