Assay Details
Target Gene Details
Entrez Gene ID: | 240476 |
Gene Name: | zinc finger protein 407 |
Gene Aliases: |
6430585N13Rik, Gm333, Gm334, Gm948, ZNF407 |
Location: |
Chr.18:84207701-84591854 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp407 | NM_001033341.2 | 2 | 853 | NP_001028513.2 |
| AK032536.1 | 2 | 854 | BAC27915.1 |
More Information
Set Membership: |
|

Genomic Map