Assay Details
Target Gene Details
Entrez Gene ID: | 240595 |
Gene Name: | potassium channel, subfamily V, member 2 |
Gene Aliases: |
KV11.1 |
Location: |
Chr.19:27322060-27337179 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnv2 | NM_183179.1 | 1 | 259 | NP_899002.1 |
| AY238939.1 | 1 | 127 | AAP70480.1 | |
| BC039042.1 | 1 | 259 | AAH39042.1 |
More Information
Set Membership: |
|

Genomic Map