Assay Details
Target Gene Details
Entrez Gene ID: | 13409 |
Gene Name: | transmembrane channel-like gene family 1 |
Gene Aliases: |
4933416G09Rik, Beethoven, Bth, CWEA1, dn |
Location: |
Chr.19:20783458-21037152 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc1 | NM_028953.2 | 21 | 2690 | NP_083229.1 |
| AF417579.1 | 21 | 2690 | AAL86400.1 | |
| AK077146.1 | 7 | 1051 | BAC36642.1 |

Genomic Map