Assay Details
Target Gene Details
Entrez Gene ID: | 17257 |
Gene Name: | methyl CpG binding protein 2 |
Gene Aliases: |
1500041B07Rik, D630021H01Rik, Mbd5, WBP10 |
Location: |
Chr.X:74026592-74085690 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mecp2 | NM_001081979.2 | 3 | 8595 | NP_001075448.1 |
| NM_010788.4 | 4 | 8719 | NP_034918.1 | |
| AF158181.1 | 4 | 8622 | AAF33024.1 | |
| AJ132922.1 | 3 | 8475 | CAB46495.1 |
More Information
Set Membership: |
|

Genomic Map