Assay Details
Target Gene Details
Entrez Gene ID: | 93843 |
Gene Name: | pregnancy upregulated non-ubiquitously expressed CaM kinase |
Gene Aliases: |
Bstk3, CaMKIbeta2, CaMKlb2, Camk1b, Punc, caMKlb1 |
Location: |
Chr.X:73655992-73660127 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnck | NM_001199351.1 | 12 | 1557 | NP_001186280.1 |
| NM_001199352.1 | 12 | 1421 | NP_001186281.1 | |
| NM_012040.3 | 12 | 1497 | NP_036170.1 | |
| AF181984.1 | 12 | 1471 | AAF29157.1 | |
| AK134665.1 | 12 | 1428 | BAE22232.1 | |
| AK158755.1 | 12 | 1422 | BAE34642.1 | |
| BC051996.1 | 12 | 1404 | AAH51996.1 | |
| BC055891.1 | 12 | 1397 | AAH55891.1 | |
| CD776443.1 | 1 | 86 |
More Information
Set Membership: |
|

Genomic Map