Assay Details
Target Gene Details
Entrez Gene ID: | 20843 |
Gene Name: | stromal antigen 2 |
Gene Aliases: |
9230105L23Rik, B230112I07Rik, SA-2, SAP2 |
Location: |
Chr.X:42149322-42277186 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stag2 | NM_001077712.2 | 1 | 175 | NP_001071180.1 |
| AK020968.1 | ||||
| AK050415.1 | ||||
| AK077318.1 | ||||
| BU920543.1 | 1 | 175 | ||
| CN669441.1 | 1 | 270 |
More Information
Set Membership: |
|

Genomic Map