Assay Details
Target Gene Details
Entrez Gene ID: | 66492 |
Gene Name: | zinc finger, matrin type 2 |
Gene Aliases: |
2610510D14Rik, 2900082I05Rik |
Location: |
Chr.18:36793923-36799660 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmat2 | NM_025594.3 | 6 | 1063 | NP_079870.1 |
| AK007521.1 | 1 | 315 | ||
| AK012091.1 | 6 | 1059 | BAB28023.1 | |
| BC024881.1 | 6 | 1063 | AAH24881.1 | |
| BC089501.1 | 6 | 1058 | AAH89501.1 |

Genomic Map