Assay Details
Target Gene Details
Entrez Gene ID: | 67116 |
Gene Name: | CUE domain containing 2 |
Gene Aliases: |
3010002G01Rik |
Location: |
Chr.19:46329812-46338660 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cuedc2 | NM_001164290.1 | 10 | 1429 | NP_001157762.1 |
| NM_001164291.1 | 9 | 1321 | NP_001157763.1 | |
| NM_001164292.1 | 9 | 1318 | NP_001157764.1 | |
| NM_001164293.1 | 9 | 1288 | NP_001157765.1 | |
| NM_001164294.1 | 9 | 1267 | NP_001157766.1 | |
| NM_001164295.1 | 9 | 1264 | NP_001157767.1 | |
| NM_024192.2 | 10 | 1420 | NP_077154.2 | |
| AK013884.1 | 9 | 1279 | BAB29035.1 |
Target Gene Details
Entrez Gene ID: | 68431 |
Gene Name: | F-box and leucine-rich repeat protein 15 |
Gene Aliases: |
0710008C12Rik, Fbxo37 |
Location: |
Chr.19:46328059-46330446 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl15 | NM_133694.2 | 4 | 1226 | NP_598455.2 |
| AK003032.1 | 4 | 949 | ||
| AK145009.1 | 4 | 1186 | ||
| AK172339.1 | 5 | 1123 | BAE42956.1 | |
| AW124512.1 | 1 | 140 | ||
| BC016499.1 | 4 | 1201 | AAH16499.1 |

Genomic Map