Assay Details
Target Gene Details
Entrez Gene ID: | 225998 |
Gene Name: | RAR-related orphan receptor beta |
Gene Aliases: |
Nr1f2, RZR-beta, RZRB, Rorbeta |
Location: |
Chr.19:18930609-19111196 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rorb | NM_001043354.2 | 10 | 2356 | NP_001036819.1 |
| NM_001289921.1 | 11 | 1875 | NP_001276850.1 | |
| NM_146095.4 | 10 | 1822 | NP_666207.3 | |
| AB258405.1 | 10 | 1983 | ||
| BC058269.1 | 9 | 1729 | AAH58269.1 |
More Information
Set Membership: |
|

Genomic Map