Assay Details
Target Gene Details
Entrez Gene ID: | 225998 |
Gene Name: | RAR-related orphan receptor beta |
Gene Aliases: |
Nr1f2, RZR-beta, RZRB, Rorbeta |
Location: |
Chr.19:18930609-19111196 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rorb | NM_001043354.2 | 4 | 1070 | NP_001036819.1 |
| NM_001289921.1 | 5 | 589 | NP_001276850.1 | |
| NM_146095.4 | 4 | 536 | NP_666207.3 | |
| AB258405.1 | 4 | 697 | ||
| AK044421.1 | 4 | 531 | ||
| AK134790.1 | 4 | 836 | ||
| BC024842.1 | 5 | 589 | ||
| BC058269.1 | 3 | 443 | AAH58269.1 | |
| DQ779923.1 | 3 | 440 | ||
| DQ779924.1 | 4 | 465 |
More Information
Set Membership: |
|

Genomic Map