Assay Details
Target Gene Details
Entrez Gene ID: | 67706 |
Gene Name: | transmembrane protein 179B |
Gene Aliases: |
0610031L02Rik, 1500003K22Rik, 1810059G22Rik |
Location: |
Chr.19:8772522-8774467 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem179b | NM_026325.3 | 5 | 738 | NP_080601.2 |
| NR_028418.1 | 3 | 415 | ||
| NR_028419.1 | 5 | 668 | ||
| AK002731.1 | 3 | 393 | ||
| AK005135.1 | 5 | 657 | ||
| AK007906.1 | 5 | 714 | BAB25340.1 | |
| AK010996.1 | 5 | 719 | BAB27315.1 | |
| BC116961.1 | ||||
| BC116963.1 |

Genomic Map