Assay Details
Target Gene Details
Entrez Gene ID: | 16825 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM2, NLI |
Location: |
Chr.19:46032589-46045516 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ldb1 | NM_001113408.1 | 11 | 1988 | NP_001106879.1 |
| NM_010697.1 | 11 | 2173 | NP_034827.1 | |
| AK048463.1 | 6 | 1207 | ||
| AK210031.1 | ||||
| BC013624.1 | 11 | 1813 | AAH13624.1 | |
| U69270.1 | 11 | 2173 | AAC52887.1 |
More Information
Set Membership: |
|

Genomic Map