Assay Details
Target Gene Details
Entrez Gene ID: | 20250 |
Gene Name: | stearoyl-Coenzyme A desaturase 2 |
Gene Aliases: |
Mir5114, Scd-2, mir-5114, swty |
Location: |
Chr.19:44293676-44306864 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scd2 | NM_009128.2 | 1 | 363 | NP_033154.2 |
| AK147406.1 | 1 | 366 | BAE27893.1 | |
| CJ110442.1 | 1 | 366 |

Genomic Map