Assay Details
Target Gene Details
Entrez Gene ID: | 20250 |
Gene Name: | stearoyl-Coenzyme A desaturase 2 |
Gene Aliases: |
Mir5114, Scd-2, mir-5114, swty |
Location: |
Chr.19:44293676-44306864 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scd2 | NM_009128.2 | 6 | 4173 | NP_033154.2 |
| AK078543.1 | 1 | 579 | ||
| AK083922.1 | BAC39066.1 | |||
| AK147406.1 | 6 | 4176 | BAE27893.1 | |
| AK159413.1 | 1 | 891 | ||
| BC040384.1 | 6 | 3748 | AAH40384.1 | |
| M26270.1 | 6 | 3761 | AAA40094.1 |

Genomic Map