Assay Details
Target Gene Details
Entrez Gene ID: | 236790 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B |
Gene Aliases: |
4930535D10Rik, 6330505F04Rik, D130066O12 |
Location: |
Chr.X:56454821-56507843 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx26b | NM_172779.4 | 1 | 404 | NP_766367.1 |
| AK031945.1 | 1 | 396 | BAC27615.1 | |
| AK083942.1 | 1 | 366 | BAC39076.1 | |
| AK160627.1 | 1 | 364 | ||
| BC051161.1 | 1 | 386 | AAH51161.1 |
More Information
Set Membership: |
|

Genomic Map