Assay Details
Target Gene Details
Entrez Gene ID: | 236790 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B |
Gene Aliases: |
4930535D10Rik, 6330505F04Rik, D130066O12 |
Location: |
Chr.X:56454821-56507843 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx26b | NM_172779.4 | 17 | 3483 | NP_766367.1 |
| AK083942.1 | 17 | 3445 | BAC39076.1 | |
| AK144026.1 | 7 | 3328 | ||
| BC019773.1 | 3 | 943 | AAH19773.1 |
More Information
Set Membership: |
|

Genomic Map