Assay Details
Target Gene Details
Entrez Gene ID: | 20977 |
Gene Name: | synaptophysin |
Gene Aliases: |
A230093K24Rik, AI848995, Syn, p38 |
Location: |
Chr.X:7638580-7653256 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syp | NM_009305.2 | 7 | 1527 | NP_033331.2 |
| AK090259.1 | 7 | 1428 | ||
| BC014823.1 | 7 | 1397 | AAH14823.1 | |
| X95818.1 | 7 | 1396 | CAA65084.1 |
More Information
Set Membership: |
|

Genomic Map