Assay Details
Target Gene Details
Entrez Gene ID: | 54130 |
Gene Name: | ARP1 actin-related protein 1A, centractin alpha |
Gene Aliases: |
Arp1, alpha-Arp1 |
Location: |
Chr.19:46376814-46395735 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr1a | NM_016860.1 | 11 | 1919 | NP_058556.1 |
| AB010297.1 | 11 | 1919 | BAA24423.1 | |
| AK010632.1 | 11 | 1916 | BAB27076.1 | |
| AK019518.1 | 10 | 1861 | ||
| AK088062.1 | 11 | 1926 | BAC40124.1 | |
| AK167450.1 | 11 | 1926 | BAE39536.1 | |
| AK168889.1 | 11 | 1923 | BAE40706.1 | |
| BC007131.1 | 11 | 1934 | AAH07131.1 |
More Information
Set Membership: |
|

Genomic Map