Assay Details
Target Gene Details
Entrez Gene ID: | 12642 |
Gene Name: | cholesterol 25-hydroxylase |
Gene Aliases: |
AI462618, m25OH |
Location: |
Chr.19:34473783-34475135 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ch25h | NM_009890.1 | 1 | 680 | NP_034020.1 |
| AF059213.1 | 1 | 680 | AAC97482.1 | |
| AK140360.1 | 1 | 698 | BAE24352.1 | |
| AK152770.1 | 1 | 689 | BAE31482.1 | |
| AK160657.1 | 1 | 694 | BAE35945.1 | |
| BC039919.1 | 1 | 713 | AAH39919.1 |
More Information
Set Membership: |
|

Genomic Map