Assay Details
Target Gene Details
Entrez Gene ID: | 12669 |
Gene Name: | cholinergic receptor, muscarinic 1, CNS |
Gene Aliases: |
Chrm-1, M1, M1R |
Location: |
Chr.19:8664005-8683606 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrm1 | NM_001112697.1 | 1 | 147 | NP_001106167.1 |
| NM_007698.3 | 1 | 147 | NP_031724.2 | |
| AJ006521.1 | CAA07082.1 | |||
| AK138282.1 | 1 | 149 | BAE23612.1 | |
| BC094242.1 | 1 | 131 | AAH94242.1 |
More Information
Set Membership: |
|

Genomic Map