Assay Details
Target Gene Details
Entrez Gene ID: | 329064 |
Gene Name: | polycystic kidney disease 2-like 1 |
Gene Aliases: |
B830002B15, BC046386, PCL, PKD2L, Pkdl, TRPP3 |
Location: |
Chr.19:44147637-44248354 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkd2l1 |
Target Gene Details
Entrez Gene ID: | 30049 |
Gene Name: | stearoyl-coenzyme A desaturase 3 |
Gene Aliases: |
4930513N16Rik |
Location: |
Chr.19:44203288-44244016 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scd3 | NM_024450.2 | 6 | 2297 | NP_077770.1 |
| AK136277.1 | 6 | 2314 | BAE22909.1 |

Genomic Map