Assay Details
Target Gene Details
Entrez Gene ID: | 19337 |
Gene Name: | RAB33A, member RAS oncogene family |
Gene Aliases: |
Rabs10 |
Location: |
Chr.X:48519285-48530240 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab33a | NM_011228.2 | 1 | 443 | NP_035358.1 |
| AB232628.1 | 1 | 127 | BAF02890.1 | |
| BC104390.1 | 1 | 191 | AAI04391.1 | |
| BC104391.1 | 1 | 191 | AAI04392.1 | |
| BX521636.1 | 1 | 413 | ||
| D83279.1 | 1 | 258 | BAA11883.1 |

Genomic Map